Learn More
Medchemexpress LLC HY-111759 10mg Medchemexpress, Jaspamycin CAS:22242-96-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11175910MG
Encompass_Preferred
Medchemexpress, HY-111759 10mg Jaspamycin CAS:22242-96-2 Jaspamycin (7-CN-7-C-Ino) is a potent activator of PKA, binding to the R site (PKAR), with an EC50 of 6.5 nM and Kd of 8 nM in Trypanosoma brucei. Jaspamycin (7-CN-7-C-Ino) does not bind with purified human PKARIα. Anti-parasite activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8953