Learn More
Cayman Chemical Jasplakinolide, 102396-24-7, 50 g
SDP
Supplier:  Cayman Chemical 1170550
Encompass_Preferred
Molecular Formula C36H45BrN4O6, Purity ≥98%, Formula Weight 709.7, Synonyms: NSC 613009; A natural macrocyclic peptide which potently inhibits the proliferation of PC3 prostate carcinoma cells (IC50 = 35 nM) by binding F-actin (KD = 15 nM); stabilizes actin filaments in vitro but disrupts actin filaments and induces irregular polymerization of monomeric actin in vivo.
Catalog No. NC0502405