Learn More
Abcam Jasplakinolide, Actin polymerization and stabilization inducer, 50UG
Catalog No. 502583360
Encompass_Preferred
MW 709.7 Da, Purity >95%. Cell-permeable actin polymerization and stabilization inducer. Potent antiproliferative activity (IC₅₀ = 15 nM). Antitumor and antifungal agent. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3360 Supplier Abcam Supplier No. AB14140950UG
MW 709.7 Da, Purity >95%. Cell-permeable actin polymerization and stabilization inducer. Potent antiproliferative activity (IC₅₀ = 15 nM). Antitumor and antifungal agent. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement