Learn More
Medchemexpress LLC HY-N0740 5mg Medchemexpress, Jatrorrhizine (chloride) CAS:6681-15-8 Purity:>98%
SDP
Catalog No. 501970307
Encompass_Preferred
Medchemexpress, HY-N0740 5mg Jatrorrhizine (chloride) CAS:6681-15-8 Jatrorrhizine chloride is a potent and orally active uptake-2 transporter inhibitor, it can be isolated from various Chinese medicinal plants. Jatrorrhizine chloride exhibits a critical neuroprotective role in H2O2-induced apoptosis via inhibition of MAPK pathway in HT22 hippocampal neurons. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0307 Supplier Medchemexpress LLC Supplier No. HYN07405MG
Medchemexpress, HY-N0740 5mg Jatrorrhizine (chloride) CAS:6681-15-8 Jatrorrhizine chloride is a potent and orally active uptake-2 transporter inhibitor, it can be isolated from various Chinese medicinal plants. Jatrorrhizine chloride exhibits a critical neuroprotective role in H2O2-induced apoptosis via inhibition of MAPK pathway in HT22 hippocampal neurons. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.