Learn More
Medchemexpress LLC Jatrorrhizine hydroxide | 483-43-2 | 355.38 | 20 MG
SDP
Catalog No. 5000416212
Encompass_Preferred
Jatrorrhizine hydroxide is an alkaloid isolated from Coptis chinensis with neuroprotective, antimicrobial, antiplasmodial, and antioxidant activities. It is a potent and orally active inhibitor of AChE (IC50=872 nM) with over >115-fold selectivity for BuChE. Jatrorrhizine hydroxide reduces the uptake of serotonin (5-HT) and norepinephrine (NE) via inhibition of uptake-2 transporters.
  • Alkaloid isolated from coptis chinensis
  • Exhibits neuroprotective activity
  • Shows antimicrobial properties
  • Possesses antiplasmodial effects
  • Offers antioxidant activities
  • Potent and orally active AChE inhibitor
  • Highly selective for BuChE over AChE
  • Reduces serotonin and norepinephrine uptake
Catalog No. 50-004-16212 Supplier Medchemexpress LLC Supplier No. HYN0749A20MG
Jatrorrhizine hydroxide is an alkaloid isolated from Coptis chinensis with neuroprotective, antimicrobial, antiplasmodial, and antioxidant activities. It is a potent and orally active inhibitor of AChE (IC50=872 nM) with over >115-fold selectivity for BuChE. Jatrorrhizine hydroxide reduces the uptake of serotonin (5-HT) and norepinephrine (NE) via inhibition of uptake-2 transporters.
  • Alkaloid isolated from coptis chinensis
  • Exhibits neuroprotective activity
  • Shows antimicrobial properties
  • Possesses antiplasmodial effects
  • Offers antioxidant activities
  • Potent and orally active AChE inhibitor
  • Highly selective for BuChE over AChE
  • Reduces serotonin and norepinephrine uptake