Learn More
Medchemexpress LLC Jatrorrhizine hydroxide | 483-43-2 | 355.38 | 50 MG
SDP
Catalog No. 5000415369
Encompass_Preferred
Jatrorrhizine hydroxide is an alkaloid isolated from *Coptis chinensis* with neuroprotective, antimicrobial, antiplasmodial, and antioxidant activities. It is a potent and orally active inhibitor of AChE (IC50=872 nM) with >115-fold selectivity for BuChE. Jatrorrhizine hydroxide reduces the uptake of serotonin (5-HT) and norepinephrine (NE) through the inhibition of uptake-2 transporters.
  • Neuroprotective activity
  • Antimicrobial activity
  • Antiplasmodial activity
  • Antioxidant activity
  • Potent and orally active inhibitor of AChE (IC50=872 nM) with high selectivity over BuChE (>115-fold)
  • Reduces uptake of serotonin (5-HT) and norepinephrine (NE) via inhibition of uptake-2 transporters
Catalog No. 50-004-15369 Supplier Medchemexpress LLC Supplier No. HYN0749A50MG
Jatrorrhizine hydroxide is an alkaloid isolated from *Coptis chinensis* with neuroprotective, antimicrobial, antiplasmodial, and antioxidant activities. It is a potent and orally active inhibitor of AChE (IC50=872 nM) with >115-fold selectivity for BuChE. Jatrorrhizine hydroxide reduces the uptake of serotonin (5-HT) and norepinephrine (NE) through the inhibition of uptake-2 transporters.
  • Neuroprotective activity
  • Antimicrobial activity
  • Antiplasmodial activity
  • Antioxidant activity
  • Potent and orally active inhibitor of AChE (IC50=872 nM) with high selectivity over BuChE (>115-fold)
  • Reduces uptake of serotonin (5-HT) and norepinephrine (NE) via inhibition of uptake-2 transporters