Learn More
Medchemexpress LLC HY-N0749A 5mg Medchemexpress, Jatrorrhizine (hydroxide) CAS:483-43-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0749A5MG
Encompass_Preferred
Medchemexpress, HY-N0749A 5mg Jatrorrhizine (hydroxide) CAS:483-43-2 Jatrorrhizine hydroxide is an alkaloid isolated from Coptis chinensis with neuroprotective, antimicrobial, antiplasmodial and antioxidant activities. Jatrorrhizine hydroxide is a potent and orally active inhibitor of AChE (IC50=872 nM) over >115-fold selectivity for BuChE. Jatrorrhizine hydroxide reduces uptake of serotonin (5-HT) and norepinephrine (NE) via inhibition of uptake-2 transporters. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2816