Learn More
Grainger RIDGID Press Jaw, Compact, 1/2 in., ASTM F 1807, 1 ea
Catalog No. 502795323
Encompass_Preferred
These compact press jaws are designed for use with 1/2 in. PEX tubing and ASTM F 1807 black copper crimp rings and insert fittings. Engineered to integrate with compact press tools, these jaws assist in creating reliable connections while significantly reducing the physical effort typically required by manual tools for larger tubing sizes.
  • Eliminates user fatigue during PEX tubing installations
  • Maintains consistent performance without requiring calibration
  • Increases the versatility of compact pressing tools
  • Compatible with 1/2 in. PEX tubing systems
  • Lifetime warranty coverage
Catalog No. 50-279-5323 Supplier Grainger Supplier No. 19U783
These compact press jaws are designed for use with 1/2 in. PEX tubing and ASTM F 1807 black copper crimp rings and insert fittings. Engineered to integrate with compact press tools, these jaws assist in creating reliable connections while significantly reducing the physical effort typically required by manual tools for larger tubing sizes.
  • Eliminates user fatigue during PEX tubing installations
  • Maintains consistent performance without requiring calibration
  • Increases the versatility of compact pressing tools
  • Compatible with 1/2 in. PEX tubing systems
  • Lifetime warranty coverage