Learn More
Grainger TB Wood's Jaw Coupling Spider, Buna-N, 80A Durometer, 1 ea
Catalog No. 502794414
Encompass_Preferred
This L190 series jaw coupling spider insert provides reliable power transmission and vibration damping in industrial coupling applications. Constructed from 80A durometer Buna-N, it is designed for compatibility with L190 L-Jaw elastomeric couplings.
  • 80A durometer Buna-N construction
  • 4.47 in. outside diameter
  • 195 N-m torque rating
  • 5,000 rpm maximum speed
  • -40°C to 212°F temperature range
  • Straight jaw design
Catalog No. 50-279-4414 Supplier Grainger Supplier No. 1A928
This L190 series jaw coupling spider insert provides reliable power transmission and vibration damping in industrial coupling applications. Constructed from 80A durometer Buna-N, it is designed for compatibility with L190 L-Jaw elastomeric couplings.
  • 80A durometer Buna-N construction
  • 4.47 in. outside diameter
  • 195 N-m torque rating
  • 5,000 rpm maximum speed
  • -40°C to 212°F temperature range
  • Straight jaw design