Learn More
Aviva Systems Biology Corp JAZF1 Antibody - N-terminal region (ARP36246_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36246_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JAZF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human JAZF1. Alternate names: DKFZp761K2222, TIP27, ZNF802ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-56089