Learn More
Electron Microscopy Sciences JB-4™ Catalyst 12 GR
SDP
Catalog No. 50980404
Encompass_Preferred
A water soluble embedding media which is based on Glycol Methacrylate (GMA) plastic embedding. It is intended for use in preparing samples for high resolution microscopes (HRLM). The catalyzed monomer acts as a dehydration and infiltration agent; therefore, complete dehydration through 100% ethanol is not necessary (although recommended for large or dense tissues). Conventional paraffin sections have a greater degree of shrinkage and produce inadequate morphology when compared to JB-4.
Catalog No. 50-980-404 Supplier Electron Microscopy Sciences Supplier No. 1427006
A water soluble embedding media which is based on Glycol Methacrylate (GMA) plastic embedding. It is intended for use in preparing samples for high resolution microscopes (HRLM). The catalyzed monomer acts as a dehydration and infiltration agent; therefore, complete dehydration through 100% ethanol is not necessary (although recommended for large or dense tissues). Conventional paraffin sections have a greater degree of shrinkage and produce inadequate morphology when compared to JB-4.