Learn More
Electron Microscopy Sciences JB-4 Plus™ Embedding Kit
SDP
Catalog No. 50980405
Encompass_Preferred
JB-4 Plus offers all of the same features as JB-4 except it produces less of an exothermic reaction than JB4 which is good for temperature sensitive tissues. J Kit consists 500 ml Solution A, 15 ml Solution B, 8 g Benzoyl Peroxide Catalyst.
Catalog No. 50-980-405 Supplier Electron Microscopy Sciences Supplier No. 1427200
JB-4 Plus offers all of the same features as JB-4 except it produces less of an exothermic reaction than JB4 which is good for temperature sensitive tissues. J Kit consists 500 ml Solution A, 15 ml Solution B, 8 g Benzoyl Peroxide Catalyst.