Learn More
Electron Microscopy Sciences JB-4 Plus™ Solution A 500 ML
SDP
Catalog No. 50980406
Encompass_Preferred
JB-4 Plus offers all of the same features as JB-4 except it produces less of an exothermic reaction than JB4 which is good for temperature sensitive tissues. J
Catalog No. 50-980-406 Supplier Electron Microscopy Sciences Supplier No. 1427201
JB-4 Plus offers all of the same features as JB-4 except it produces less of an exothermic reaction than JB4 which is good for temperature sensitive tissues. J