Learn More
Medchemexpress LLC JB-95 acetate | 98.4% | 1971.41 (free base) | 10 MG
SDP
Catalog No. 5000398377
Encompass_Preferred
JB-95 acetate is a β-hairpin macrocyclic peptide that demonstrates potent antimicrobial activity against *Escherichia coli*. It selectively disrupts the outer membrane of *E. coli* but not the inner membrane.
  • β-hairpin macrocyclic peptide
  • Potent antimicrobial activity against *Escherichia coli*
  • Selectively disrupts the outer membrane of *E. coli*
Catalog No. 50-003-98377 Supplier Medchemexpress LLC Supplier No. HYP5753A10MG
Add to Cart
Add to Cart
JB-95 acetate is a β-hairpin macrocyclic peptide that demonstrates potent antimicrobial activity against *Escherichia coli*. It selectively disrupts the outer membrane of *E. coli* but not the inner membrane.
  • β-hairpin macrocyclic peptide
  • Potent antimicrobial activity against *Escherichia coli*
  • Selectively disrupts the outer membrane of *E. coli*