Learn More
Medchemexpress LLC JB-95 acetate | 98.4% | 1971.41 (free base) | 5 MG
SDP
Catalog No. 5000399126
Encompass_Preferred
JB-95 acetate, a β-hairpin macrocyclic peptide, exhibits potent antimicrobial activity against Escherichia coli. It can selectively disrupt the outer membrane but not the inner membrane of E. coli.
  • Potent antimicrobial activity
  • Exhibits activity against Escherichia coli
  • Selectively disrupts the outer membrane of E. coli
  • Does not disrupt the inner membrane of E. coli
  • β-hairpin macrocyclic peptide
Catalog No. 50-003-99126 Supplier Medchemexpress LLC Supplier No. HYP5753A5MG
JB-95 acetate, a β-hairpin macrocyclic peptide, exhibits potent antimicrobial activity against Escherichia coli. It can selectively disrupt the outer membrane but not the inner membrane of E. coli.
  • Potent antimicrobial activity
  • Exhibits activity against Escherichia coli
  • Selectively disrupts the outer membrane of E. coli
  • Does not disrupt the inner membrane of E. coli
  • β-hairpin macrocyclic peptide