Learn More
Grainger Epoxy Adhesive, Tube, 5.0 oz, Gray
Catalog No. NC1784177
Encompass_Preferred
J-B Weld™ Professional not only creates a strong, permanent bond on virtually any clean, dry surface, but can also be drilled, sanded, filled, tapped, machined, and painted once it's fully cured, for a piece that is just about as workable as a solid. Use on wood, metal, tile, ceramic, jewelry, glass, plastic, and more. Petroleum-, chemical-, and acid-resistant.
Catalog No. NC1784177 Supplier Grainger Supplier No. 2UV88
J-B Weld™ Professional not only creates a strong, permanent bond on virtually any clean, dry surface, but can also be drilled, sanded, filled, tapped, machined, and painted once it's fully cured, for a piece that is just about as workable as a solid. Use on wood, metal, tile, ceramic, jewelry, glass, plastic, and more. Petroleum-, chemical-, and acid-resistant.