Learn More
Medchemexpress LLC JBI-589 | 2308504-22-3 | C29H28FN5O | 1 ML
SDP
Catalog No. 5000373189
Encompass_Preferred
JBI-589 is a non-covalent, orally bioavailable, isoform-selective inhibitor of PAD4 (Protein Arginine Deiminase 4). It is suitable for use in cancer research, as it has been shown to reduce CXCR2 expression and block neutrophil chemotaxis, while also reducing primary tumor and metastases. This compound can enhance the anti-tumor effects of checkpoint inhibitors.
  • Significantly downregulates CXCR2 expression on neutrophils
  • Reduces CXCL1-induced migration
  • Inhibits citrullination of histone H3 in induced mouse neutrophils
  • Significantly inhibits primary tumor growth in mouse models
  • Leads to a significant downregulation of CXCR2 receptor expression in Ly6G cells
Catalog No. 50-003-73189 Supplier Medchemexpress LLC Supplier No. HY15345010MM/1ML
JBI-589 is a non-covalent, orally bioavailable, isoform-selective inhibitor of PAD4 (Protein Arginine Deiminase 4). It is suitable for use in cancer research, as it has been shown to reduce CXCR2 expression and block neutrophil chemotaxis, while also reducing primary tumor and metastases. This compound can enhance the anti-tumor effects of checkpoint inhibitors.
  • Significantly downregulates CXCR2 expression on neutrophils
  • Reduces CXCL1-induced migration
  • Inhibits citrullination of histone H3 in induced mouse neutrophils
  • Significantly inhibits primary tumor growth in mouse models
  • Leads to a significant downregulation of CXCR2 receptor expression in Ly6G cells