Learn More
MSC JBO Thread Ring Gage, NO GO, UNF Nr.6, 40 Pitch, 2A Tolerance
Catalog No. 5000531239
Encompass
This thread ring gage is designed for checking UNF (Unified National Fine Thread) with a nominal diameter of Nr.6 and a pitch of 40. It features a NO GO design and a 2A tolerance, ensuring accurate thread inspection. This uncoated gage is suitable for various applications requiring precise thread verification.
  • No GO thread ring gage (AR)
  • Unified National Fine Thread (UNF) form
  • Nominal diameter of Nr.6
  • 40 pitch
  • 2A tolerance
  • Uncoated finish
Catalog No. 50-005-31239 Supplier MSC Supplier No. 55322838
This thread ring gage is designed for checking UNF (Unified National Fine Thread) with a nominal diameter of Nr.6 and a pitch of 40. It features a NO GO design and a 2A tolerance, ensuring accurate thread inspection. This uncoated gage is suitable for various applications requiring precise thread verification.
  • No GO thread ring gage (AR)
  • Unified National Fine Thread (UNF) form
  • Nominal diameter of Nr.6
  • 40 pitch
  • 2A tolerance
  • Uncoated finish