Learn More
Medchemexpress LLC HY-112584 100mg Medchemexpress, JBSNF-000088 CAS:7150-23-4 Purity:>98%
SDP
Catalog No. 501969107
Encompass_Preferred
Medchemexpress, HY-112584 100mg JBSNF-000088 CAS:7150-23-4 JBSNF-000088 (6-Methoxynicotinamide), a analog of nicotinamide (NA), is a potent Nicotinamide N-methyltransferase (NNMT) inhibitor with IC50s of 1.8 µM, 2.8 µM, and 5.0 µM for human NNMT, monkey NNMT and mouse NNMT, respectively. JBSNF-000088 inhibits NNMT activity, reduces MNA levels and drives insulin sensitization, glucose modulation and body weight reduction in animal models of metabolic disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9107 Supplier Medchemexpress LLC Supplier No. HY112584100MG
Medchemexpress, HY-112584 100mg JBSNF-000088 CAS:7150-23-4 JBSNF-000088 (6-Methoxynicotinamide), a analog of nicotinamide (NA), is a potent Nicotinamide N-methyltransferase (NNMT) inhibitor with IC50s of 1.8 µM, 2.8 µM, and 5.0 µM for human NNMT, monkey NNMT and mouse NNMT, respectively. JBSNF-000088 inhibits NNMT activity, reduces MNA levels and drives insulin sensitization, glucose modulation and body weight reduction in animal models of metabolic disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.