Learn More
Medchemexpress LLC HY-15534 10mg , JC-1 CAS:3520-43-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1553410MG
Encompass_Preferred
HY-15534 10mg JC-1 CAS: 3520-43-2 JC-1 (CBIC2) is a fluorescent lipophilic carbocyanine dye used to measure mitochondrial membrane potential. JC-1 forms complexes known as J-aggregates at high ΔΨm. Aggregates of JC-1 emit an orange-red fluorescence (Ex/Em=585/590 nm). While in cells with low ΔΨm, JC-1 remains in the monomeric form. JC-1 monomers emit a green fluorescence (Ex/Em=510/527 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0167