Learn More
AdipoGen JC-1
SDP
Supplier:  AdipoGen CDXT0046M010
Encompass_Preferred
Chemical. CAS 3520-43-2. Formula C25H27Cl4IN4. MW 652.23. Synthetic. A dual-emission potential-sensitive probe that can be used to measure mitochondrial membrane potential. JC-1 is a green-fluorescent monomer at low membrane potential. At higher potentials, JC-1 forms red-fluorescent "J-aggregates," which exhibit broad excitation and very narrow emission spectra. The ratio of red to green fluorescence of JC-1 is dependent only on membrane potential, and not influenced by mitochondrial size, shape, or density.
Catalog No. 50-256-0032