Learn More
Medchemexpress LLC HY-15534 5mg Medchemexpress, JC-1 CAS:3520-43-2 Purity:>98%
SDP
Catalog No. 502029137
Encompass_Preferred
Medchemexpress, HY-15534 5mg JC-1 CAS:3520-43-2 JC-1 (CBIC2) is a fluorescent lipophilic carbocyanine dye used to measure mitochondrial membrane potential. JC-1 forms complexes known as J-aggregates at high ΔΨm. Aggregates of JC-1 emit an orange-red fluorescence (Ex/Em=585/590 nm). While in cells with low ΔΨm, JC-1 remains in the monomeric form. JC-1 monomers emit a green fluorescence (Ex/Em=510/527 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9137 Supplier Medchemexpress LLC Supplier No. HY155345MG
Medchemexpress, HY-15534 5mg JC-1 CAS:3520-43-2 JC-1 (CBIC2) is a fluorescent lipophilic carbocyanine dye used to measure mitochondrial membrane potential. JC-1 forms complexes known as J-aggregates at high ΔΨm. Aggregates of JC-1 emit an orange-red fluorescence (Ex/Em=585/590 nm). While in cells with low ΔΨm, JC-1 remains in the monomeric form. JC-1 monomers emit a green fluorescence (Ex/Em=510/527 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.