Learn More
Abcam JC-1, Mitochondrial membrane potential dye, 5MG
Catalog No. 502583363
Encompass_Preferred
MW 652.2 Da, Purity >95%. Mitochondrial membrane-potential dye. Exhibits potential-dependent accumulation in mitochondria. Fluorescence emission shift from green (~529 nm) to red (~590 nm). Depolarization is indicated by a decrease in the red/green fluorescence intensity ratio.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3363 Supplier Abcam Supplier No. AB1413875MG
MW 652.2 Da, Purity >95%. Mitochondrial membrane-potential dye. Exhibits potential-dependent accumulation in mitochondria. Fluorescence emission shift from green (~529 nm) to red (~590 nm). Depolarization is indicated by a decrease in the red/green fluorescence intensity ratio.
The product is subject to the following: Abcam Restricted Use Statement