Learn More
Novus Biologicals™ JC-1 mitochondrial membrane potential dependent fluorescent dye

Description
- Purity:>98% (HPLC); NMR (conforms)
- CAS No.: 3520-43-2
- Molecular Formula: C25H27Cl4IN4
- Molecular Weight:652.2
- Source:Synthetic
- Appearance:Red powder
- PubChem ID: 5492929
Specifications
Specifications
| Format | Red powder |
| Quantity | 1 mg |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.