Learn More
Apexbio Technology LLC JC-10 Mitochondrial Membrane Potential Assay Kit(Synonyms: JC-10 Assay Kit, JC-10 MMP Assay Kit, JC-10 mitochondrial dye assay), 100T.
SDP
Catalog No. 5000061375
Encompass_Preferred
The JC-10 Mitochondrial Membrane Potential Assay Kit utilizes the fluorescent probe JC-10 to assess mitochondrial membrane potential ( m) a critical marker for mitochondrial function and early apoptosis detection JC-10 selectively accumulates in mitochondria with intact membrane potential forming red-fluorescent aggregates Upon loss of m associated with apoptosis or mitochondrial dysfunction JC-10 disperses into monomeric forms emitting green fluorescence The mitochondrial membrane potential is therefore indicated by changes in the ratio between red and green fluorescence intensity This assay is suitable for studies involving cultured cells isolated mitochondria or tissue samples and is commonly employed in apoptosis research and mitochondrial functional analysis
Catalog No. 50-000-61375 Supplier Apexbio Technology LLC Supplier No. K2001100
The JC-10 Mitochondrial Membrane Potential Assay Kit utilizes the fluorescent probe JC-10 to assess mitochondrial membrane potential ( m) a critical marker for mitochondrial function and early apoptosis detection JC-10 selectively accumulates in mitochondria with intact membrane potential forming red-fluorescent aggregates Upon loss of m associated with apoptosis or mitochondrial dysfunction JC-10 disperses into monomeric forms emitting green fluorescence The mitochondrial membrane potential is therefore indicated by changes in the ratio between red and green fluorescence intensity This assay is suitable for studies involving cultured cells isolated mitochondria or tissue samples and is commonly employed in apoptosis research and mitochondrial functional analysis