Learn More
Enzo Life Sciences JC-10 (ultra pure), (5mg)
SDP
Catalog No. NC0591190
Encompass_Preferred
Mitochondria dye . Alternative Name: Enhanced JC-1. MW: ~600. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Use/Stability: Stable for at least one year after receipt when stored as recommended. Long Term Storage: -20°C
Catalog No. NC0591190 Supplier Enzo Life Sciences Supplier No. ENZ52305
Mitochondria dye . Alternative Name: Enhanced JC-1. MW: ~600. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Use/Stability: Stable for at least one year after receipt when stored as recommended. Long Term Storage: -20°C