Learn More
Medchemexpress LLC JC-171 | 2112809-98-8 | MFCD32708504 | 98.4% | 384.83 g/mol | C16H17ClN2O5S | 5 MG
SDP
Catalog No. 5000227773
Encompass_Preferred
JC-171 is a selective NLRP3 inflammasome inhibitor that blocks LPS/ATP-induced interleukin-1β (IL-1β) release from macrophages (IC50 8.45 μM). It is used for in vitro studies of inflammasome activation and for in vivo models of inflammasome-driven inflammation such as experimental autoimmune encephalomyelitis.
  • Selective NLRP3 inflammasome inhibition
  • Effective in macrophage IL-1β release assays (IC50 8.45 μM)
  • Validated in vivo activity in experimental autoimmune encephalomyelitis
  • High purity solid for research use (≈98.4%)
  • Stable as powder under recommended storage conditions
  • Available in small research quantities (e.g., 5 mg)
Catalog No. 50-002-27773 Supplier Medchemexpress LLC Supplier No. HY1174325MG
JC-171 is a selective NLRP3 inflammasome inhibitor that blocks LPS/ATP-induced interleukin-1β (IL-1β) release from macrophages (IC50 8.45 μM). It is used for in vitro studies of inflammasome activation and for in vivo models of inflammasome-driven inflammation such as experimental autoimmune encephalomyelitis.
  • Selective NLRP3 inflammasome inhibition
  • Effective in macrophage IL-1β release assays (IC50 8.45 μM)
  • Validated in vivo activity in experimental autoimmune encephalomyelitis
  • High purity solid for research use (≈98.4%)
  • Stable as powder under recommended storage conditions
  • Available in small research quantities (e.g., 5 mg)