Learn More
ATCC JC; Mammary Adenocarcinoma; Mouse (Mus musculus)
Supplier:  ATCC CRL2116
Encompass
JC are epithelial-like cells that were isolated from the breasts of a mouse with adenocarcinoma. It has applications in cancer research.
Catalog No. 50-238-3244