Learn More
Molecular Dimensions JCSG-PLUS 10ML
SDP
Catalog No. NC0085789
Encompass_Preferred
Protein Crystallography. JCSG-plus screen (96 x 10mL) is the screen of choice for initial screening experiments.The most complete sparse matrix screen available today. Screen classic PEG and salt conditions more efficiently. Modern precipitants. Range of salts.Broad pH range.
Catalog No. NC0085789 Supplier Molecular Dimensions Supplier No. MD137
Protein Crystallography. JCSG-plus screen (96 x 10mL) is the screen of choice for initial screening experiments.The most complete sparse matrix screen available today. Screen classic PEG and salt conditions more efficiently. Modern precipitants. Range of salts.Broad pH range.