Learn More
TARGETMOL CHEMICALS INC JD-5037 10MG
SDP
Catalog No. 502579881
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JD-5037 is a novel peripherally restricted CB1R antagonist with an IC50 of 1.5 nM. purity: 98%
Catalog No. 50-257-9881 Supplier TARGETMOL CHEMICALS INC Supplier No. T445310MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JD-5037 is a novel peripherally restricted CB1R antagonist with an IC50 of 1.5 nM. purity: 98%