Learn More
Medchemexpress LLC HY-18697 100mg Medchemexpress, JD-5037 CAS:1392116-14-1 Purity:98%
SDP
Catalog No. 501638807
Encompass_Preferred
Medchemexpress,HY-18697 100mg JD-5037 CAS:1392116-14-1 Formula:C27H27Cl2N5O3S JD-5037 is a novel, peripherally restricted CB1R antagonist with an IC50 of 1.5 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8807 Supplier Medchemexpress LLC Supplier No. HY18697100MG
Medchemexpress,HY-18697 100mg JD-5037 CAS:1392116-14-1 Formula:C27H27Cl2N5O3S JD-5037 is a novel, peripherally restricted CB1R antagonist with an IC50 of 1.5 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.