Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec JD500 Tray Bars
GSA_VASDP

Catalog No. 22444180 Shop All Mopec Products
Change view
Click to view available options
Weight (English):
2200 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
22-444-180 2200 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-180 Supplier Mopec Supplier No. JD500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Consists of two heavy-duty bars along with nylon straps that securely attach to the hand slots of the body trays for easy elevation. Mopec™ JD500 Tray Bars is to be used in conjunction with the JD Series Cadaver Lifts.

  • 2 heavy duty support bars
  • 4 nylon straps: 1 x 36 in. basket
  • Basket weight rated at 2,200 Pounds
  • For use with JD Series cadaver lift

Warranty

1 Year Warranty

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Cadaver Lift
Description 1 in. x 36 in. Nylon Straps
Product Line Lifts
Weight (English) 2200 lb.
Weight (Metric) 99.79 Kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.