Learn More
Thwing Albert Instrument Co JDC-3-10 SAMPLE CUTTER W/O SHIELDS
SDP
Catalog No. NC3135945
Encompass
JDC-3-10 Precision Sample Cutter to cut sample strips 3" wide and up to 10 inches long ** NOTincluding safety shields OR platforms**.
Catalog No. NC3135945 Supplier Thwing Albert Instrument Co Supplier No. 000992068
Add to Cart
Add to Cart
Request a Quote
JDC-3-10 Precision Sample Cutter to cut sample strips 3" wide and up to 10 inches long ** NOTincluding safety shields OR platforms**.