Learn More
JDM LAB SOLUTIONS LLC JDM Lab Solutions. Unique Multi-Head™ for general purpose vortexing PLUS holds up to eight microtubes.Powerful motor for INSTANT vortexing. Variable speed from 200 to 3200 rpm
Catalog No. NC4045553
Encompass
JDM Lab Solutions. Unique Multi-Head™ for general purpose vortexing PLUS holds up to eight microtubes.Powerful motor for INSTANT vortexing. Variable speed from 200 to 3200 rpm
Catalog No. NC4045553 Supplier JDM LAB SOLUTIONS LLC Supplier No. JDMV1005
JDM Lab Solutions. Unique Multi-Head™ for general purpose vortexing PLUS holds up to eight microtubes.Powerful motor for INSTANT vortexing. Variable speed from 200 to 3200 rpm