Learn More
JDM LAB SOLUTIONS LLC JDM Lab SolutionsReversible top platform, cup or flat-dimpled surface. Powerful motor for INSTANT vortexing. Variable speed from 200 to 3200 rpm. Wide variety of accessories, for tubes up to 50ml
Catalog No. NC4045554
Encompass
JDM Lab SolutionsReversible top platform, cup or flat-dimpled surface. Powerful motor for INSTANT vortexing. Variable speed from 200 to 3200 rpm. Wide variety of accessories, for tubes up to 50ml
Catalog No. NC4045554 Supplier JDM LAB SOLUTIONS LLC Supplier No. JDMV1003
JDM Lab SolutionsReversible top platform, cup or flat-dimpled surface. Powerful motor for INSTANT vortexing. Variable speed from 200 to 3200 rpm. Wide variety of accessories, for tubes up to 50ml