Learn More
Selleck Chemical LLC Opnurasib-E1056-5MG
SDP
Supplier:  Selleck Chemical LLC E10565MG
Encompass_Preferred
Opnurasib (JDQ-443 NVP-JDQ443) a potent and selective orally bioavailable covalent inhibitor of GDP-bound KRASG12C with an IC50 of 0 012 M for inhibition of c-Raf recruitment
Catalog No. 50-238-9793