Learn More
Medchemexpress LLC HY-10487 10mg , JDTic (dihydrochloride) CAS:785835-79-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1048710MG
Encompass_Preferred
HY-10487 10mg JDTic (dihydrochloride) CAS: 785835-79-2 JDTic (dihydrochloride) is a potent antagonist of kappa-opioid receptors (KOR), blocking the κ-agonist U50, 488-induced antinociception. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9444