Learn More
Medchemexpress LLC HY-10487 5mg Medchemexpress, JDTic (dihydrochloride) CAS:785835-79-2 Purity:>98%
SDP
Catalog No. 501968235
Encompass_Preferred
Medchemexpress, HY-10487 5mg JDTic (diHCl) CAS:785835-79-2 JDTic (dihydrochloride) is a potent antagonist of kappa-opioid receptors (KOR), blocking the κ-agonist U50, 488-induced antinociception. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8235 Supplier Medchemexpress LLC Supplier No. HY104875MG
Medchemexpress, HY-10487 5mg JDTic (diHCl) CAS:785835-79-2 JDTic (dihydrochloride) is a potent antagonist of kappa-opioid receptors (KOR), blocking the κ-agonist U50, 488-induced antinociception. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.