Learn More
Medchemexpress LLC JDTic dihydrochloride | 785835-79-2 | MFCD22419279 | 98.0% | 538.55 | C28H41Cl2N3O3 | 25 MG
SDP
Catalog No. 5000243422
Encompass_Preferred
JDTic dihydrochloride is the dihydrochloride salt of JDTic, a potent and selective kappa-opioid receptor (KOR) antagonist used as a research reagent to inhibit κ-agonist (e.g., U50,488)-induced antinociception and to study KOR signaling. It is supplied as a characterized solid for laboratory assays and is not for human use.
  • Potent kappa-opioid receptor (KOR) antagonist used in pharmacological research.
  • Blocks κ-agonist U50,488-induced antinociception in model systems.
  • High purity (98.03%) suitable for research applications.
  • White to off-white solid appearance.
  • Dissolves in DMSO (100 mg/mL) and H2O (50 mg/mL); ultrasonic assistance recommended.
  • Store sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or patient use.
Catalog No. 50-002-43422 Supplier Medchemexpress LLC Supplier No. HY1048725MG
JDTic dihydrochloride is the dihydrochloride salt of JDTic, a potent and selective kappa-opioid receptor (KOR) antagonist used as a research reagent to inhibit κ-agonist (e.g., U50,488)-induced antinociception and to study KOR signaling. It is supplied as a characterized solid for laboratory assays and is not for human use.
  • Potent kappa-opioid receptor (KOR) antagonist used in pharmacological research.
  • Blocks κ-agonist U50,488-induced antinociception in model systems.
  • High purity (98.03%) suitable for research applications.
  • White to off-white solid appearance.
  • Dissolves in DMSO (100 mg/mL) and H2O (50 mg/mL); ultrasonic assistance recommended.
  • Store sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or patient use.