Learn More
Medchemexpress LLC JDTic dihydrochloride | 785835-79-2 | MFCD22419279 | 98.0% | 538.55 g·mol⁻¹ | C28H41Cl2N3O3 | 1 ML
SDP
Catalog No. 5000244055
Encompass_Preferred
JDTic dihydrochloride is the dihydrochloride salt of JDTic, a selective kappa-opioid receptor (KOR) antagonist used in research to block κ-agonist-induced antinociception. Supplied as a concentrated solution for in vitro pharmacology and receptor signaling studies.
  • Selective kappa-opioid receptor antagonist suitable for pharmacology research.
  • Provided as a 10 mM solution in a 1 mL vial.
  • High purity appropriate for research applications.
  • Soluble in DMSO and water for assay preparation.
  • Commonly used for receptor antagonism and KOR signaling studies.
Catalog No. 50-002-44055 Supplier Medchemexpress LLC Supplier No. HY1048710MM 1ML
JDTic dihydrochloride is the dihydrochloride salt of JDTic, a selective kappa-opioid receptor (KOR) antagonist used in research to block κ-agonist-induced antinociception. Supplied as a concentrated solution for in vitro pharmacology and receptor signaling studies.
  • Selective kappa-opioid receptor antagonist suitable for pharmacology research.
  • Provided as a 10 mM solution in a 1 mL vial.
  • High purity appropriate for research applications.
  • Soluble in DMSO and water for assay preparation.
  • Commonly used for receptor antagonism and KOR signaling studies.