Learn More
Medchemexpress LLC JDTic dihydrochloride | 785835-79-2 | MFCD22419279 | 98.0% | 538.55 g/mol | C28H41Cl2N3O3 | 50 MG
SDP
Catalog No. 5000244445
Encompass_Preferred
JDTic dihydrochloride is a research reagent that acts as a potent kappa-opioid receptor (KOR) antagonist. It is used in pharmacology and neuroscience studies to block κ-agonist-induced antinociception and to probe KOR-mediated signaling pathways. The compound is supplied as a white to off-white solid with reported high purity.
  • Potent kappa-opioid receptor antagonist.
  • Useful for studies of receptor pharmacology and antinociception.
  • High purity (98.0%).
  • Solid, white to off-white, soluble in DMSO and water with ultrasonic assistance.
  • Stable as a solid at 4°C when sealed; solutions stored frozen for long-term use.
  • Molecular weight 538.55 g/mol; formula C28H41Cl2N3O3.
Catalog No. 50-002-44445 Supplier Medchemexpress LLC Supplier No. HY1048750MG
JDTic dihydrochloride is a research reagent that acts as a potent kappa-opioid receptor (KOR) antagonist. It is used in pharmacology and neuroscience studies to block κ-agonist-induced antinociception and to probe KOR-mediated signaling pathways. The compound is supplied as a white to off-white solid with reported high purity.
  • Potent kappa-opioid receptor antagonist.
  • Useful for studies of receptor pharmacology and antinociception.
  • High purity (98.0%).
  • Solid, white to off-white, soluble in DMSO and water with ultrasonic assistance.
  • Stable as a solid at 4°C when sealed; solutions stored frozen for long-term use.
  • Molecular weight 538.55 g/mol; formula C28H41Cl2N3O3.