Learn More
Sklar Instruments Jeans Dissecting Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel. Sklar's Jeans Dissecting Forceps are ideal for separating and dissecting tissue during surgical procedures. EA/1.
SDP
Catalog No. 501437427 Shop All Sklar Instruments Products
Encompass_Preferred
Jeans Dissecting Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel. Sklar's Jeans Dissecting Forceps are ideal for separating and dissecting tissue during surgical procedures. They have a light-weight fenstrated thumb handle, and teeth at the tips.
This item is not returnable. View return policy
Catalog No. 50-143-7427 Supplier Sklar Instruments Supplier No. 192181
Jeans Dissecting Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel. Sklar's Jeans Dissecting Forceps are ideal for separating and dissecting tissue during surgical procedures. They have a light-weight fenstrated thumb handle, and teeth at the tips.