Learn More
ATCC JEG-3; Choriocarcinoma; Human (Homo sapiens)
Catalog No. 502383096
Encompass
JEG-3 is a hypertriploid, clonally-derived, human cell line with epithelial morphology that was isolated from the Woods strain of the Erwin-Turner tumor. This cell line is tumorigenic, capable of forming malignant tumors consistent with choriocarcionoma. Genes expressed: human chorionic gonadotropin (hCG); human chorionic somatomammotropin (placental lactogen); progesterone.
Catalog No. 50-238-3096 Supplier ATCC Supplier No. HTB36
JEG-3 is a hypertriploid, clonally-derived, human cell line with epithelial morphology that was isolated from the Woods strain of the Erwin-Turner tumor. This cell line is tumorigenic, capable of forming malignant tumors consistent with choriocarcionoma. Genes expressed: human chorionic gonadotropin (hCG); human chorionic somatomammotropin (placental lactogen); progesterone.