Learn More
Jeio Tech Inc Jeio Tech™ BS2-30, 55L Shaking Heating Bath (230V, 60Hz), Precision Temperature Control, Adjustable Agitation, Digital Interface, Stainless Steel Tank, Safety Protection
SDP
Catalog No. NC3789804
Encompass_Preferred
Jeio Tech BS2-30 Shaking Heating Bath, Precision Temperature Control and Agitation. Featuring PID temperature control (Amb +5°C to 100°C) and adjustable shaking speeds (30-200 rpm), it ensures uniform heating and consistent mixing. The durable stainless steel inner bath (width 22.2" × length 12.6" × depth 7.6") supports a wide range of applications including incubation and cell culture. A touchscreen controller enables easy operation with programmable timer and calibration (3-point temp, 1-point shaking). The gable cover accommodates up to 2.8L flasks while preventing condensation drip. Safety features include low-water alarm (visual/audio), deviation alarms, and overcurrent protection. Additional features include drain port, spring wire rack for various vessels, torque hinge lid, and optional remote monitoring.
Catalog No. NC3789804 Supplier Jeio Tech Inc Supplier No. AAH44305K
Jeio Tech BS2-30 Shaking Heating Bath, Precision Temperature Control and Agitation. Featuring PID temperature control (Amb +5°C to 100°C) and adjustable shaking speeds (30-200 rpm), it ensures uniform heating and consistent mixing. The durable stainless steel inner bath (width 22.2" × length 12.6" × depth 7.6") supports a wide range of applications including incubation and cell culture. A touchscreen controller enables easy operation with programmable timer and calibration (3-point temp, 1-point shaking). The gable cover accommodates up to 2.8L flasks while preventing condensation drip. Safety features include low-water alarm (visual/audio), deviation alarms, and overcurrent protection. Additional features include drain port, spring wire rack for various vessels, torque hinge lid, and optional remote monitoring.