Learn More
Jeio Tech Inc Jeio Tech™ IB4-05V, 60L Advanced Forced Convection Incubator (120V, 60Hz), PID Temperature Control, Temperature Ramp, Data Logging (RS-232/USB), Uniform Heating
SDP
Catalog No. NC3965854
Encompass_Preferred
Jeio Tech IB4-05V Advanced Incubator, Forced Convection with PID Control delivers precise, uniform temperature performance for biological and clinical applications. With excellent stability (±0.15°C fluctuation) and uniformity (±0.41°C variation at 37°C), it ensures consistent conditions for culture and testing. Rapid recovery restores 37°C within 5 minutes after a 30-second door opening. The advanced system features adjustable fan speed, temperature ramping, 3-point calibration, and a 5-inch touchscreen controller with real-time monitoring. RS-232/USB connectivity supports data logging and remote access. Built with a durable chamber and adjustable shelving, it offers reliable performance for research and pharma labs. Inner chamber: W 14.96" × D 12.99" × H 18.9".
Catalog No. NC3965854 Supplier Jeio Tech Inc Supplier No. AAH21663U
Jeio Tech IB4-05V Advanced Incubator, Forced Convection with PID Control delivers precise, uniform temperature performance for biological and clinical applications. With excellent stability (±0.15°C fluctuation) and uniformity (±0.41°C variation at 37°C), it ensures consistent conditions for culture and testing. Rapid recovery restores 37°C within 5 minutes after a 30-second door opening. The advanced system features adjustable fan speed, temperature ramping, 3-point calibration, and a 5-inch touchscreen controller with real-time monitoring. RS-232/USB connectivity supports data logging and remote access. Built with a durable chamber and adjustable shelving, it offers reliable performance for research and pharma labs. Inner chamber: W 14.96" × D 12.99" × H 18.9".