Learn More
Jeio Tech Inc Jeio Tech™ IB4-15AV, 150L Advanced Air Jacket Incubator (120V, 60Hz), PID Temperature Control, Uniform Heating, Temperature Ramping, Data Logging (RS-232/USB)
SDP
Catalog No. NC3965869
Encompass_Preferred
Jeio Tech IB4-15AV Air-Jacket Advanced Incubator delivers precise and uniform temperature control using a high-performance air-jacket heating system, ideal for sensitive biological and clinical applications. The air-jacket design ensures gentle and stable temperature distribution, reducing the risk of thermal fluctuations and protecting delicate cultures. Equipped with an intuitive digital PID controller, it provides excellent stability (±0.15°C fluctuation) and uniformity (±0.55°C variation at 37°C), while the advanced model includes features such as fan speed control, programmable temperature ramping, real-time graphical monitoring, and RS-232/USB communication ports for data logging and remote access. With a durable corrosion-resistant inner chamber ( W 21.62" × D 14.8" × H 27.95"), adjustable shelving, and built-in safety protections, the IB4-A is the optimal choice for reliable incubation in microbiology, biotechnology, and pharmaceutical laboratories.
Catalog No. NC3965869 Supplier Jeio Tech Inc Supplier No. AAH21783U
Jeio Tech IB4-15AV Air-Jacket Advanced Incubator delivers precise and uniform temperature control using a high-performance air-jacket heating system, ideal for sensitive biological and clinical applications. The air-jacket design ensures gentle and stable temperature distribution, reducing the risk of thermal fluctuations and protecting delicate cultures. Equipped with an intuitive digital PID controller, it provides excellent stability (±0.15°C fluctuation) and uniformity (±0.55°C variation at 37°C), while the advanced model includes features such as fan speed control, programmable temperature ramping, real-time graphical monitoring, and RS-232/USB communication ports for data logging and remote access. With a durable corrosion-resistant inner chamber ( W 21.62" × D 14.8" × H 27.95"), adjustable shelving, and built-in safety protections, the IB4-A is the optimal choice for reliable incubation in microbiology, biotechnology, and pharmaceutical laboratories.