Learn More
Jeio Tech Inc Jeio Tech™ MS-32M Multi Position Magnetic Stirrer (3 Rows x 2 Columns, 100-240V, 50/60Hz), Feedback Speed Control, BLDC Motor, Strong Magnetic Coupling, Timer, Spill-Proof Design, Quiet Mixing
SDP
Catalog No. NC3965951
Encompass_Preferred
Jeio Tech MS-32M Multi-Position Magnetic Stirrer, High-Throughput Mixing with Precise Speed Control delivers efficient and flexible performance for demanding lab workflows. Featuring feedback control and a wide speed range (30-2000 rpm), it ensures accurate, consistent stirring across all positions. The durable BLDC motor provides quiet, maintenance-free operation, while strong permanent magnets enable stable magnetic coupling. Flexible control options allow independent point, row, or column operation to suit various applications. The smooth start/stop function minimizes splashing for safer handling, and the spill-proof design protects internal components. A built-in timer supports programmable mixing, and an optional silicone pad enhances vessel stability. With optimized spacing (117 mm width, 90 mm depth), the MS-32M is ideal for reliable, high-throughput laboratory mixing.
Catalog No. NC3965951 Supplier Jeio Tech Inc Supplier No. AAH332215U
Jeio Tech MS-32M Multi-Position Magnetic Stirrer, High-Throughput Mixing with Precise Speed Control delivers efficient and flexible performance for demanding lab workflows. Featuring feedback control and a wide speed range (30-2000 rpm), it ensures accurate, consistent stirring across all positions. The durable BLDC motor provides quiet, maintenance-free operation, while strong permanent magnets enable stable magnetic coupling. Flexible control options allow independent point, row, or column operation to suit various applications. The smooth start/stop function minimizes splashing for safer handling, and the spill-proof design protects internal components. A built-in timer supports programmable mixing, and an optional silicone pad enhances vessel stability. With optimized spacing (117 mm width, 90 mm depth), the MS-32M is ideal for reliable, high-throughput laboratory mixing.