Learn More
Jeio Tech Inc Jeio Tech™ MS-33M Multi Position Magnetic Stirrer (3 Rows x 3 Columns, 100-240V, 50/60Hz), Feedback Speed Control, BLDC Motor, Strong Magnetic Coupling, Timer, Spill-Proof Design, Quiet Mixing
SDP
Catalog No. NC3965954
Encompass_Preferred
Jeio Tech MS-33M Multi-Position Magnetic Stirrer, High-Throughput Mixing with Precise Speed Control delivers efficient and flexible performance for demanding lab workflows. Featuring feedback control and a wide speed range (30-2000 rpm), it ensures accurate, consistent stirring across all positions. The durable BLDC motor provides quiet, maintenance-free operation, while strong permanent magnets enable stable magnetic coupling. Flexible control options allow independent point, row, or column operation to suit various applications. The smooth start/stop function minimizes splashing for safer handling, and the spill-proof design protects internal components. A built-in timer supports programmable mixing, and an optional silicone pad enhances vessel stability. With optimized spacing (117 mm width, 90 mm depth), the MS-33M is ideal for reliable, high-throughput laboratory mixing.
Catalog No. NC3965954 Supplier Jeio Tech Inc Supplier No. AAH332315U
Jeio Tech MS-33M Multi-Position Magnetic Stirrer, High-Throughput Mixing with Precise Speed Control delivers efficient and flexible performance for demanding lab workflows. Featuring feedback control and a wide speed range (30-2000 rpm), it ensures accurate, consistent stirring across all positions. The durable BLDC motor provides quiet, maintenance-free operation, while strong permanent magnets enable stable magnetic coupling. Flexible control options allow independent point, row, or column operation to suit various applications. The smooth start/stop function minimizes splashing for safer handling, and the spill-proof design protects internal components. A built-in timer supports programmable mixing, and an optional silicone pad enhances vessel stability. With optimized spacing (117 mm width, 90 mm depth), the MS-33M is ideal for reliable, high-throughput laboratory mixing.