Learn More
Jeio Tech Inc Jeio Tech™ OF4-03VW, 26L Advanced Forced Convection (Mechanical) Oven with Window (120V, 60Hz), PID Temperature Control, Fast Uniform Heating, Temperature Ramping, Data Logging (RS-232/USB)
SDP
Catalog No. NC3965927
Encompass_Preferred
Jeio Tech OF4-03VW Advanced Windowed Forced Convection (Mechanical) Oven is designed for fast, uniform, and efficient thermal processing. Featuring a powerful forced-air circulation system, the OF4 series ensures rapid temperature recovery and uniform heat distribution, making it ideal for drying, curing, heating, and stability testing of various samples. PID control system delivers excellent stability (±0.3°C fluctuation) and uniformity (±1.4°C variation) across a temperature range of Amb.+10°C to 250°C. The advanced model includes temperature ramping, and RS-232/USB communication ports for data logging and external control, auto-tuning, 3-point calibration, 5-inch LCD touchscreen with real-time graphs, and safety protections. With reliable performance and user-friendly operation, the OF4 series is ideal for use in research, industrial, pharmaceutical, and quality control laboratories. Inner chamber: W 11.8 x D 8.3 x H 15.8 inches.
Catalog No. NC3965927 Supplier Jeio Tech Inc Supplier No. AAH12816U
Jeio Tech OF4-03VW Advanced Windowed Forced Convection (Mechanical) Oven is designed for fast, uniform, and efficient thermal processing. Featuring a powerful forced-air circulation system, the OF4 series ensures rapid temperature recovery and uniform heat distribution, making it ideal for drying, curing, heating, and stability testing of various samples. PID control system delivers excellent stability (±0.3°C fluctuation) and uniformity (±1.4°C variation) across a temperature range of Amb.+10°C to 250°C. The advanced model includes temperature ramping, and RS-232/USB communication ports for data logging and external control, auto-tuning, 3-point calibration, 5-inch LCD touchscreen with real-time graphs, and safety protections. With reliable performance and user-friendly operation, the OF4 series is ideal for use in research, industrial, pharmaceutical, and quality control laboratories. Inner chamber: W 11.8 x D 8.3 x H 15.8 inches.