Learn More
Jeio Tech Inc Jeio Tech™ OF4-05V, 61L Advanced Forced Convection (Mechanical) Oven (120V, 60Hz), PID Temperature Control, Fast Uniform Heating, Temperature Ramping, Data Logging (RS-232/USB)
SDP
Catalog No. NC3965917
Encompass_Preferred
Jeio Tech OF4-05V Advanced Forced Convection (Mechanical) Oven is designed for fast, uniform, and efficient thermal processing. Featuring a powerful forced-air circulation system, the OF4 series ensures rapid temperature recovery and uniform heat distribution, making it ideal for drying, curing, heat-treating, and stability testing of various samples. The PID control system delivers excellent stability (±0.3°C fluctuation) and uniformity (±1.9°C variation) across a temperature range of Amb.+10°C to 250°C. The advanced model includes temperature ramping, and RS-232/USB communication ports for data logging and external control, auto-tuning, 3-point calibration, 5-inch LCD touchscreen with real-time graphs, and safety protections. With reliable performance and user-friendly operation, the OF4 series is ideal for use in research, industrial, pharmaceutical, and quality control laboratories. Inner chanber: W 15 x D 13 x H 18.9 inches.
Catalog No. NC3965917 Supplier Jeio Tech Inc Supplier No. AAH12726U
Jeio Tech OF4-05V Advanced Forced Convection (Mechanical) Oven is designed for fast, uniform, and efficient thermal processing. Featuring a powerful forced-air circulation system, the OF4 series ensures rapid temperature recovery and uniform heat distribution, making it ideal for drying, curing, heat-treating, and stability testing of various samples. The PID control system delivers excellent stability (±0.3°C fluctuation) and uniformity (±1.9°C variation) across a temperature range of Amb.+10°C to 250°C. The advanced model includes temperature ramping, and RS-232/USB communication ports for data logging and external control, auto-tuning, 3-point calibration, 5-inch LCD touchscreen with real-time graphs, and safety protections. With reliable performance and user-friendly operation, the OF4 series is ideal for use in research, industrial, pharmaceutical, and quality control laboratories. Inner chanber: W 15 x D 13 x H 18.9 inches.